Tisk…

3,5-Dichloro-2-hydroxybenzenesulphonyl chloride, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL146110-25GEA 0 CZK
SIAL146110-25G
3,5-Dichloro-2-hydroxybenzenesulphonyl chloride, Sigma-Aldrich®
3,5-Dichloro-2-hydroxybenzenesulphonyl chloride
Vzorec: C₆H₃Cl₃O₃S
MW: 261,51
MDL Number: MFCD00007432
CAS číslo: 23378-88-3
EINECS: 245-619-9
UN: 3261
ADR: 8,I

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR