Tisk…

5-(Trifluoromethoxy)salicylaldehyde, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL342157-500MGEA 0 CZK
SIAL342157-500MG
5-(Trifluoromethoxy)salicylaldehyde, Sigma-Aldrich®
5-(Trifluoromethoxy)salicylaldehyde
Vzorec: C₈H₅F₃O₃
MW: 206,12
MDL Number: MFCD00075249
CAS číslo: 93249-62-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR