Tisk…

2-Hydroxy-5-methylisophthalaldehyde, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL393851-1GEA 0 CZK
SIAL393851-1G
2-Hydroxy-5-methylisophthalaldehyde, Sigma-Aldrich®
2-Hydroxy-5-methylisophthalaldehyde
Vzorec: C₉H₈O₃
MW: 164,1600 g/mol
MDL Number: MFCD00016596
CAS číslo: 7310-95-4
EINECS: 230-768-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR