Isobornyl acetate
Dodavatel: EHRENSTORFER GMBH
EHERC14385000EA
1560
CZK
EHERC14385000
Isobornyl acetate
Isobornyl acetate
Vzorec:
C₁₂H₂₀O₂ MW: 196,29 g/mol |
MDL Number:
MFCD00135943 CAS číslo: 125-12-2 EINECS: 204-727-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...