Tisk…

2'-Methoxyacetanilide, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL485063-100GEA 1250 CZK
SIAL485063-100G
2'-Methoxyacetanilide, Sigma-Aldrich®
2'-Methoxyacetanilide
Vzorec: C₉H₁₁NO₂
MW: 165,1891 g/mol
MDL Number: MFCD00026117
CAS číslo: 93-26-5
EINECS: 202-233-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR