Tisk…

3-Chloropropyl acetate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

SIAL166561-25GEA 1080 CZK
SIAL166561-25G
3-Chloropropyl acetate, Sigma-Aldrich®
3-Chloropropyl acetate
Vzorec: C₅H₉ClO₂
MW: 136,58 g/mol
MDL Number: MFCD00001000
CAS číslo: 628-09-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR