Tisk…
Ethyl arachidate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
SIALA9010-10GEA 22130 CZK
SIALA9010-10G SIALA9010-1G
Ethyl arachidate, Sigma-Aldrich®
Ethyl arachidate
Vzorec: C₂₂H₄₄O₂
MW: 340,5800 g/mol
MDL Number: MFCD00056233
CAS číslo: 18281-05-5