Tisk…

5-Bromo-2,4-dimethoxybenzaldehyde, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL292516-5GEA 0 CZK
SIAL292516-5G
5-Bromo-2,4-dimethoxybenzaldehyde, Sigma-Aldrich®
5-Bromo-2,4-dimethoxybenzaldehyde
Vzorec: C₉H₉BrO₃
MW: 245,0700 g/mol
MDL Number: MFCD00010863
CAS číslo: 130333-46-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR