Tisk…

5-Iodosalicylaldehyde, Sigma-Aldrich®

Dodavatel: Merck

SIAL524328-1GEA 0 CZK
SIAL524328-1G
5-Iodosalicylaldehyde, Sigma-Aldrich®
5-Iodosalicylaldehyde
Vzorec: C₇H₅IO₂
MW: 248,0200 g/mol
MDL Number: MFCD00723501
CAS číslo: 1761-62-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR