1,2,4,5-Tetrakis(bromomethyl)benzene, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
SIAL236616-5GEA
6140
CZK
SIAL236616-5G
1,2,4,5-Tetrakis(bromomethyl)benzene, Sigma-Aldrich®
1,2,4,5-Tetrakis(bromomethyl)benzene
Vzorec:
C₁₀H₁₀Br₄ MW: 449,8 g/mol |
MDL Number:
MFCD00009664 CAS číslo: 15442-91-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...