Tetrakis(dimethylamino)zirconium(IV), Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
SIAL669016-25GEA
0
CZK
SIAL669016-25G
Tetrakis(dimethylamino)zirconium(IV), Sigma-Aldrich®
Tetrakis(dimethylamino)zirconium(IV)
Vzorec:
C₈H₂₄N₄Zr MW: 267,527 g/mol |
MDL Number:
MFCD00239502 CAS číslo: 19756-04-8 UN: 3396 ADR: 4.3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...