Tetrakis(ethylmethylamino)zirconium(IV), Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
SIAL553131-5GEA
16940
CZK
SIAL553131-5G
Tetrakis(ethylmethylamino)zirconium(IV), Sigma-Aldrich®
Tetrakis(ethylmethylamino)zirconium(IV)
Vzorec:
C₁₂H₃₂N₄Zr |
MDL Number:
MFCD03427131 CAS číslo: 175923-04-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...