Tisk…

4-Cyano-3-fluorobenzaldehyde, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL682454-500MGEA 0 CZK
SIAL682454-500MG
4-Cyano-3-fluorobenzaldehyde, Sigma-Aldrich®
4-Cyano-3-fluorobenzaldehyde
Vzorec: C₈H₄FNO
MW: 149,1200 g/mol
MDL Number: MFCD06738989
CAS číslo: 101048-76-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR