Tisk…

2,4-Dimethoxy-6-methylbenzaldehyde, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL667749-5GEA 0 CZK
SIAL667749-5G
2,4-Dimethoxy-6-methylbenzaldehyde, Sigma-Aldrich®
2,4-Dimethoxy-6-methylbenzaldehyde
Vzorec: C₁₀H₁₂O₃
MDL Number: MFCD00033851
CAS číslo: 7149-90-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR