Order Entry
Trichloroethylene
Trichloroethylene
  LACH20058AT0M10001
 :  
undefined
Trichloroethylene
  LACH20058AT0M10001
 :  
  1 * 1 L
  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
 :
 :12,09769
 :86.7 °C (1013 hPa)
 :–73 °C
 :1.46 g/cm³ (20 °C)
 :Ambient
 :201-167-4
 :1710
 :6.1,III
 :131.39 g/mol
 :GHS08
 :GHS07
 :79-01-6
 :ClCH=CCl₂
 :MFCD00000838