Tisk…
Storage Boxes with Lid
Dodavatel: Semadeni
216-3146EA 390 CZK
SEMA3360
Storage Boxes with Lid
These storage boxes with dividers are made of polystyrene (PS).
  • Fitted with a lid and two partitions with adjustable dividers
  • These dividers can be placed in 19 different positions