Formalin acetic acid, Q Path® histological fixative
Dodavatel: VWR Q-Path Chemicals
|
Nebezpečí
|
Q path®
11699025.EA
871
CZK
11699025.
Formalin acetic acid, Q Path® histological fixative
Formalin acetic acid
Formalin acetic acid ready to use fixative for tissue samples
Vzorec:
CH₂O Storage Temperature: Ambient |
MDL Number:
MFCD00003274 CAS číslo: 50-00-0 EINECS: 200-001-8 |
Výsledky specifikačního testu
Appearance | Colourless liquid |
Density (20/4) | 1.012 - 1.022 |
pH (20°C) | 2.1 - 2.5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...