Tisk…

(S)-(+)-Glycidyl methyl ether, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL454699-1GEA 4090 CZK
SIAL454699-1G
(S)-(+)-Glycidyl methyl ether, Sigma-Aldrich®
(S)-(+)-Glycidyl methyl ether
Vzorec: C₄H₈O₂
MDL Number: MFCD00273366
CAS číslo: 64491-68-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR