(1S)-(+)-Camphor-10-sulphonamide, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
SIAL376612-1GEA
1070
CZK
SIAL376612-1G
(1S)-(+)-Camphor-10-sulphonamide, Sigma-Aldrich®
(1S)-(+)-Camphor-10-sulphonamide
Vzorec:
C₁₀H₁₇NO₃S MW: 231,32 g/mol |
MDL Number:
MFCD00075611 CAS číslo: 60933-63-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...