2-Aminoimidazole hemisulphate
MOLE29958183-25GEA
0
CZK
MOLE29958183-25G
2-Aminoimidazole hemisulphate
2-Aminoimidazole hemisulphate
Vzorec:
C₆H₁₂N₆O₄S MW: 264,26 g/mol |
MDL Number:
MFCD00013162 CAS číslo: 1450-93-7 EINECS: 215-918-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...