Tisk…

2-Butyl-5-chloroimidazole-4-carbaldehyde, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL559881-25GEA 0 CZK
SIAL559881-25G
2-Butyl-5-chloroimidazole-4-carbaldehyde, Sigma-Aldrich®
2-Butyl-5-chloroimidazole-4-carbaldehyde
Vzorec: C₈H₁₁ClN₂O
MDL Number: MFCD01934396
CAS číslo: 83857-96-9
EINECS: 410-260-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR