Histamine dihydrochloride
Dodavatel: EHRENSTORFER GMBH
EHERC14213050EA
3270
CZK
EHERC14213050
Histamine dihydrochloride
Histamine dihydrochloride
Vzorec:
C₅H₁₁Cl₂N₃ MW: 184,07 g/mol |
MDL Number:
MFCD00012703 CAS číslo: 56-92-8 EINECS: 200-298-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...