4-Formyl-1-naphthaleneboronic acid ≥95%
|
Varování
|
H53143.MDEA
16920
CZK
H53143.MD
4-Formyl-1-naphthaleneboronic acid ≥95%
4-Formyl-1-naphthaleneboronic acid
Vzorec:
C₁₁H₉BO₃ MW: 200,00 g/mol Bod tání: 230 °C Storage Temperature: Ambient |
MDL Number:
MFCD01632205 CAS číslo: 332398-52-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...