Tisk…

Eucalyptus oil globulus, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ300

SIALW246611-5KG-KEA 0 CZK
SIALW246611-5KG-K SIALW246611-10KG-K
Eucalyptus oil globulus, Sigma-Aldrich®
Eucalyptus oil globulus
MDL Number: MFCD00146880
CAS číslo: 85203-56-1
EINECS: 286-249-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR