Tisk…

Olive oil, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

SIAL75343-1LEA 4510 CZK
SIAL75343-1L
Olive oil, Sigma-Aldrich®
Olive oil
MDL Number: MFCD00131764
CAS číslo: 8001-25-0
EINECS: 232-277-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR