Dimethyl-trans-cyclohexane-1,4-dicarboxylate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
SIAL366684-1GEA
1410
CZK
SIAL366684-1G
Dimethyl-trans-cyclohexane-1,4-dicarboxylate, Sigma-Aldrich®
Dimethyl-trans-cyclohexane-1,4-dicarboxylate
Vzorec:
C₁₀H₁₆O₄ MW: 200,23 g/mol |
MDL Number:
MFCD00063917 CAS číslo: 3399-22-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...