3-(4-Morpholino)propyl isothiocyanate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
SIAL560464-1GEA
723
CZK
SIAL560464-1G
3-(4-Morpholino)propyl isothiocyanate, Sigma-Aldrich®
3-(4-Morpholino)propyl isothiocyanate
Vzorec:
C₈H₁₄N₂OS MW: 186,28 g/mol |
MDL Number:
MFCD00041222 CAS číslo: 32813-50-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...