4-(4-Chlorophenyl)-3-thiosemicarbazide ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
L12657.06EA
5010
CZK
L12657.06
4-(4-Chlorophenyl)-3-thiosemicarbazide ≥97%
4-(4-Chlorophenyl)-3-thiosemicarbazide
Vzorec:
C₇H₈ClN₃S Bod tání: ∼ 180 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00041295 CAS číslo: 22814-92-2 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...