Tisk…

2-Chloro-5-(trifluoromethyl)benzyl alcohol, Sigma-Aldrich®

Dodavatel: Merck

SIAL714607-5GEA 0 CZK
SIAL714607-5G
2-Chloro-5-(trifluoromethyl)benzyl alcohol, Sigma-Aldrich®
2-Chloro-5-(trifluoromethyl)benzyl alcohol
Vzorec: C₈H₆ClF₃O
MW: 210,58
MDL Number: MFCD01631469
CAS číslo: 64372-62-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR