5-Isoquinolineboronic acid
Synonyms:
Isoquinoline-5-boronic acid
, Isoquinolin-5-ylboronic acid
MOLEM87104849EA
0
CZK
MOLEM87104849
5-Isoquinolineboronic acid
5-Isoquinolineboronic acid
Vzorec:
C₉H₈BNO₂ MW: 172,98 g/mol |
MDL Number:
MFCD03839356 CAS číslo: 371766-08-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...