Tisk…

(R)-(-)-2-Hexyl isothiocyanate 95%

Dodavatel: Thermo Scientific
Nebezpečí

L20066.03EA 2610 CZK
L20066.03
(R)-(-)-2-Hexyl isothiocyanate 95%
(R)-(-)-2-Hexyl isothiocyanate
Vzorec: C₇H₁₃NS
Hustota: 0,91
Storage Temperature: Refrigerator
MDL Number: MFCD05664053
CAS číslo: 737000-95-2
EINECS: 000-000-0
UN: 2922
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR