4-(2,4-Dimethylphenyl)-3-thiosemicarbazide ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
L10338.03EA
869
CZK
L10338.03
4-(2,4-Dimethylphenyl)-3-thiosemicarbazide ≥98%
4-(2,4-Dimethylphenyl)-3-thiosemicarbazide
Vzorec:
C₉H₁₃N₃S MW: 195,29 g/mol Bod tání: 150…152 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041279 CAS číslo: 66298-09-7 EINECS: 000-000-0 UN: 2811 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...