4-Mercaptobenzoic acid
MOLE49383024-5GEA
4970
CZK
MOLE49383024-5G
4-Mercaptobenzoic acid
4-Mercaptobenzoic acid
Vzorec:
C₇H₆O₂S MW: 154,1863 g/mol |
MDL Number:
MFCD00016617 CAS číslo: 1074-36-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...