Tisk…
Cut resistant gloves, HyFlex® 11-500
Dodavatel: Ansell
HyFlex®
ANSE11-500/8EA 0 CZK
ANSE11-500/8 ANSE11-500/7
Cut resistant gloves, HyFlex® 11-500
Rukavice
These gloves with Lycra®/nylon/Kevlar® lining and nitrile palm coating are designed for dry or slightly oily applications where workers need light cut protection, and in some cases, isolation from heat or flying molten metal debris.
  • Excellent dexterity and breathability
  • Offering optimal comfort and a sure grip: versatile choice for optimal productivity and safety
  • Silicone-free
  • Anti-static according to EN 1149
They are well suited for a broad range of assembly applications, such as the assembly of brown goods, white goods, engine parts, small components and oil-coated pieces.
Certifikace: EN 388:2016 (3231B), EN 407 (X1XXXX), EN 1149; Cat. III