forensicGEM® DNA extraction kits have been developed to advance capabilities in forensic profiling by offering higher throughput levels, in manual or automated workflows, as well as a reduction in sample processing time and consumable consumption.
forensicGEM® Cigarette kit utilises careful optimization of the forensicGEM Saliva method to generate buffers and methods that gently release DNA from cigarette butts while leaving the tars, phenolics and other inhibitors behind.
forensicGEM® Cigarette kit utilises careful optimisation of the forensicGEM Saliva method to generate buffers and methods that gently release DNA from cigarette butts while leaving the tars, phenolics and other inhibitors behind.