Tisk…
Flame-retardant rain jackets, high visibility, Andilly 9728
Dodavatel: Sioen
SIOE9728/FY1/2XLEA 7200 CZK
SIOE9728/FY1/2XL SIOE9728/FY1/L SIOE9728/FY1/M SIOE9728/FY1/3XL SIOE9728/FY1/1XL SIOE9728/FC1/S SIOE9728/FC1/3XL SIOE9728/FC1/2XL SIOE9728/FY1/S SIOE9728/FC1/1XL SIOE9728/FC1/L SIOE9728/FC1/M
Flame-retardant rain jackets, high visibility, Andilly 9728
Anti-static, unlined jacket made Flexothane® Anti-static fabric (100% FR polyester knitting with FR PU coating). Different linings or fleeces can be zipped in.
  • Straight collar with foldaway hood
  • Zip closure under double flap with press studs
  • Two inset pockets
  • Kimono sleeves, sleeve narrowing by press studs
  • High frequency welded seams
  • Flame retardant reflective tape (50 mm)
Back length: 85 cm
Weight: 200 g/m²
Certifikace: EN ISO 14116 (Index 1/25H/40), EN 1149-5, EN 13034 (Type PB (6)), EN ISO 20471 (Class 3), EN 343 + A1 (3-1), EN ISO 13688