3-(Methoxycarbonyl)phenylboronic acid pinacol ester, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
SIAL594652-1GEA
3750
CZK
SIAL594652-1G
3-(Methoxycarbonyl)phenylboronic acid pinacol ester, Sigma-Aldrich®
3-(Methoxycarbonyl)phenylboronic acid pinacol ester
Vzorec:
C₁₄H₁₉BO₄ MW: 262,1093 g/mol |
MDL Number:
MFCD03789258 CAS číslo: 480425-35-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...