5-(tert-Butylcarbamoyl)-2-fluorophenylboronic acid ≥97%
|
Varování
|
H53080.03EA
15100
CZK
H53080.03
5-(tert-Butylcarbamoyl)-2-fluorophenylboronic acid ≥97%
5-(tert-Butylcarbamoyl)-2-fluorophenylboronic acid
Vzorec:
C₁₁H₁₅BFNO₃ MW: 239,05 g/mol Bod tání: 142…144 °C Storage Temperature: Ambient |
MDL Number:
MFCD08436054 CAS číslo: 874289-51-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...