(S)-(-)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid
MOLE42731182-5GEA
0
CZK
MOLE42731182-5G
(S)-(-)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid
(S)-(-)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid
Vzorec:
C₁₀H₁₁NO₂ MW: 177,20 g/mol |
MDL Number:
MFCD00144533 CAS číslo: 74163-81-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...