Tisk…

(S)-(-)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL421626-5GEA 2370 CZK
SIAL421626-5G
(S)-(-)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid, Sigma-Aldrich®
(S)-(-)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid
Vzorec: C₁₀H₁₁NO₂
MW: 177,20 g/mol
MDL Number: MFCD00144533
CAS číslo: 74163-81-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR