Tisk…

Ammonium-[¹⁵N] nitrate (60% ¹⁵N), Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

SIAL609455-5GEA 13810 CZK
SIAL609455-5G
Ammonium-[¹⁵N] nitrate (60% ¹⁵N), Sigma-Aldrich®
Ammonium-[¹⁵N] nitrate
Vzorec: H₄N₂O₃
MDL Number: MFCD00064510
CAS číslo: 31432-48-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR