Tisk…

2-(4-Bromophenyl)-6-iodoimidazo[1,2-a]pyridine, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL658995-5GEA 0 CZK
SIAL658995-5G SIAL658995-1G
2-(4-Bromophenyl)-6-iodoimidazo[1,2-a]pyridine, Sigma-Aldrich®
2-(4-Bromophenyl)-6-iodoimidazo[1,2-a]pyridine
Vzorec: C₁₃H₈BrIN₂
MW: 399,03
MDL Number: MFCD00444892
CAS číslo: 118000-66-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR