Tisk…

Iodomethane[¹²C] (99.9% ¹³C ) stabilizovaný, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí


Stabilization: Stabilised with copper
M-Clarity™ quality level = MQ200

SIAL606731-5GEA 6790 CZK
SIAL606731-5G
Iodomethane[¹²C] (99.9% ¹³C ) stabilizovaný, Sigma-Aldrich®
Iodomethane[¹²C]

Depleted of ¹³C

Vzorec: CH₃I
MW: 141,93 g/mol
MDL Number: MFCD00198114
CAS číslo: 1173018-72-8
UN: 2644
ADR: ,

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR