Buffers, potassium phosphate buffer (0,5 mol/L) solutions
Supplier: Cayman Chemical
Certificates
About this item
Product Information
- Catalog #Supplier #pH valuePack typePkAvailabilityPrice
- Potassium phosphate buffer solution (0,5 mol/l)Potassium phosphate buffer solution (0,5 mol/l)CAYM600207-250600207-2506,0Glass bottle250 mlLog in for availability1 * 250 mL1 840,00 KčSupplier Article Number: 600207-250Local Article Number: CAYM600207-250
- Potassium phosphate buffer solution (0,5 mol/l)Potassium phosphate buffer solution (0,5 mol/l)CAYM600208-250600208-2507,0Glass bottle250 mlLog in for availability1 * 250 mL1 840,00 KčSupplier Article Number: 600208-250Local Article Number: CAYM600208-250
- Potassium phosphate buffer solution (0,5 mol/l)Potassium phosphate buffer solution (0,5 mol/l)CAYM600209-250600209-2508,0Glass bottle250 mlLog in for availability1 * 250 mL1 840,00 KčSupplier Article Number: 600209-250Local Article Number: CAYM600209-250
- Potassium phosphate buffer solution (0,5 mol/l)Potassium phosphate buffer solution (0,5 mol/l)CAYM600210-250600210-2509,0Glass bottle250 mlLog in for availability1 * 250 mL1 750,00 KčSupplier Article Number: 600210-250Local Article Number: CAYM600210-250
- Potassium phosphate buffer solution (0,5 mol/l)Potassium phosphate buffer solution (0,5 mol/l)CAYM600207-500600207-5006,0Glass bottle500 mlLog in for availability1 * 500 mL3 470,00 KčSupplier Article Number: 600207-500Local Article Number: CAYM600207-500
- Potassium phosphate buffer solution (0,5 mol/l)Potassium phosphate buffer solution (0,5 mol/l)CAYM600208-500600208-5007,0Glass bottle500 mlLog in for availability1 * 500 mL3 470,00 KčSupplier Article Number: 600208-500Local Article Number: CAYM600208-500
- Potassium phosphate buffer solution (0,5 mol/l)Potassium phosphate buffer solution (0,5 mol/l)CAYM600209-500600209-5008,0Glass bottle500 mlLog in for availability1 * 500 mL3 470,00 KčSupplier Article Number: 600209-500Local Article Number: CAYM600209-500
- Potassium phosphate buffer solution (0,5 mol/l)Potassium phosphate buffer solution (0,5 mol/l)CAYM600210-500600210-5009,0Glass bottle500 mLLog in for availability1 * 500 mL3 310,00 KčSupplier Article Number: 600210-500Local Article Number: CAYM600210-500
- Potassium phosphate buffer solution (0,5 mol/l)Potassium phosphate buffer solution (0,5 mol/l)CAYM600207-1600207-16,0Glass bottle1 LLog in for availability1 * 1 L6 320,00 KčSupplier Article Number: 600207-1Local Article Number: CAYM600207-1
- Potassium phosphate buffer solution (0,5 mol/l)Potassium phosphate buffer solution (0,5 mol/l)CAYM600208-1600208-17,0Glass bottle1 LLog in for availability1 * 1 L6 500,00 KčSupplier Article Number: 600208-1Local Article Number: CAYM600208-1
- Potassium phosphate buffer solution (0,5 mol/l)Potassium phosphate buffer solution (0,5 mol/l)CAYM600209-1600209-18,0Glass bottle1 LLog in for availability1 * 1 L6 500,00 KčSupplier Article Number: 600209-1Local Article Number: CAYM600209-1
- Potassium phosphate buffer solution (0,5 mol/l)Potassium phosphate buffer solution (0,5 mol/l)CAYM600210-1600210-19,0Glass bottle1 LLog in for availability1 * 1 L6 190,00 KčSupplier Article Number: 600210-1Local Article Number: CAYM600210-1