Tisk…

Cyclopropylmagnesium bromide 0.5 mol/l in THF, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL526797-100MLEA 0 CZK
SIAL526797-100ML
Cyclopropylmagnesium bromide 0.5 mol/l in THF, Sigma-Aldrich®
Cyclopropylmagnesium bromide
Vzorec: C₃H₅BrMg
MW: 145,28
MDL Number: MFCD00672094
CAS číslo: 23719-80-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR