Tisk…

Diisopropylzinc 1 mol/l in toluene, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL568112-25MLEA 0 CZK
SIAL568112-25ML SIAL568112-5ML
Diisopropylzinc 1 mol/l in toluene, Sigma-Aldrich®
Diisopropylzinc
Vzorec: C₆H₁₄Zn
MDL Number: MFCD01862461
CAS číslo: 625-81-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR