Tisk…

Dimethylzinc 1 mol/l in heptane, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

SIAL417246-50MLEA 0 CZK
SIAL417246-50ML
Dimethylzinc 1 mol/l in heptane, Sigma-Aldrich®
Dimethylzinc
Vzorec: C₂H₆Zn
MW: 95,478 g/mol
MDL Number: MFCD00014854
CAS číslo: 544-97-8
EINECS: 208-884-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR