Tropylium tetrafluoroborate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
SIAL164623-10GEA
25630
CZK
SIAL164623-10G
Tropylium tetrafluoroborate, Sigma-Aldrich®
Tropylium tetrafluoroborate
Vzorec:
C₇H₇BF₄ MW: 177,94 g/mol |
MDL Number:
MFCD00013402 CAS číslo: 27081-10-3 EINECS: 248-214-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...