Tisk…

5-Benzyl-1H-tetrazole ≥99%

Dodavatel: Thermo Scientific

H50675.03EA 2550 CZK
H50675.03
5-Benzyl-1H-tetrazole ≥99%
5-Benzyl-1H-tetrazole
Vzorec: C₈H₈N₄
MW: 160,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00068730
CAS číslo: 18489-25-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR