Tisk…

2-(4-Boc-piperazino)-2-(4-fluorophenyl)acetic acid, Sigma-Aldrich®

Dodavatel: Merck

SIAL44264-1G-FEA 0 CZK
SIAL44264-1G-F SIAL44264-250MG-F
2-(4-Boc-piperazino)-2-(4-fluorophenyl)acetic acid, Sigma-Aldrich®
2-(4-Boc-piperazino)-2-(4-fluorophenyl)acetic acid
Vzorec: C₁₇H₂₃FN₂O₄
MDL Number: MFCD01862498
CAS číslo: 868151-70-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR